Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

59280-U

SSI Fittings Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

packaging

pkg of 1 ea

General description

Save time when looking for the fitting you need. This kit contains everything you need to connect most columns to a 1/16 in. tubing system, regardless of the column manufacturer. It is designed for easy storage and easy parts identification -- everything is clearly labeled to help you quickly locate what you need.

The kit contains:
  • SSI ferrules (5)
  • E-Z Grip gland nuts (3)
  • SSI gland nuts (5)
  • CPI fittings, KEL-F (2)
  • CPI ferrules (5)
  • SSI coupling, 0.10 in. I.D. (1)
  • CPI gland nuts (5)
  • SSI to CPI couplings (2)
  • E-Z Grip ferrules (3)

Legal Information

SSI is a trademark of Scientific Systems, Inc.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service