Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

59249-U

Supelco

SUPELCOGEL 8Ca (9 µm) HPLC Columns

L × I.D. 25 cm × 4.6 mm

Synonym(s):

SUPELCOGEL 8% Crosslinked HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

Product Name

SUPELCOGEL 8Ca, 8% Crosslinked HPLC Column, 9 μm particle size, L × I.D. 25 cm × 4.6 mm

material

stainless steel column

agency

suitable for USP L19

product line

SUPELCOGEL

feature

endcapped: no

manufacturer/tradename

SUPELCOGEL

packaging

1 ea of

parameter

400 bar pressure (5801 psi)
85 °C max. temp.

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.6 mm

matrix

sulfonated polystyrene/divinylbenzene, spherical
polymeric

particle size

9 μm

separation technique

ligand exchange

Looking for similar products? Visit Product Comparison Guide

General description

SUPELCOGEL 8 Column Series (manufactured with 8% XL PS-DVB):
The majority of columns on the market for the analysis of carbohydrates, organic acids, and alcohols, such as Aminex®, utilize an 8% XL PS-DVB. The 8% polymer offers excellent separation of many common samples.
Advantages: 8% polymers combine durability and good resolution. Due to higher cross-linking than 6% XL polymers, columns packed with 8% polymer can be used at higher flow rates therefore decreasing analysis times.
Disadvantages: 8% polymers are limited in separating carbohydrates larger than trisaccharides due to lower MW exclusion limits. Compared to the 6% XL polymer, for some types of carbohydrate and organic acid mixtures, the 8% shows less resolution due to selectivity differences.

Molecular Weight Exclusion Limit: 1200
Minimum Wet Capacity meq/ml: 1.7 - 1.9
Flow Rate Recommendation: 0.4 - 1.2 ml/min

Legal Information

Aminex is a registered trademark of Bio-Rad Laboratories, Inc.
SUPELCOGEL is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service