Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5919221

Supelco

SUPELCOSIL ABZ+Plus HPLC Column

3 μm particle size, L × I.D. 5 cm × 2.1 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

agency

suitable for USP L60

feature

endcapped

extent of labeling

12.0% carbon loading

parameter

≤70 °C temp. range

technique(s)

HPLC: suitable

L × I.D.

5 cm × 2.1 mm

surface area

170 m2/g

surface coverage

surface coverage 3.4 μmol/m2

matrix

silica gel, spherical particle platform

matrix active group

amide, alkyl phase

particle size

3 μm

pore size

120 Å

pH range

2-7.5

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

SUPELCOSIL ABZ+Plus columns offer both high deactivation and unique selectivity. Deactivated silica particles of very narrow particle size distribution ensure high efficiency with low back pressure. After bonding and endcapping reactions, the ABZ+Plus phase effectively shields unreacted silanol groups on the silica, preventing them from interacting with most analytes, and provides symmetric peaks regardless of an analyte′s functionality. The phase also allows you to use low ionic strength buffers without having to add an ion-suppressing modifier. ABZ+Plus enables you to use simple mobile phases when analyzing the most difficult compounds; acids, strongly basic compounds, and zwitterions.
suitable for L60 per USP

Features and Benefits

• High efficiency for polar, nonpolar, and charged analytes
• Symmetric peaks for the most reactive compounds
• Unique selectivity for polar and charged compounds

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhongping John Lin et al.
Journal of pharmaceutical and biomedical analysis, 37(4), 757-762 (2005-03-31)
A highly sensitive method was developed and validated for determining the free fraction of flunarizine in human plasma. Equilibrium dialysis was used for the separation of free (unbound) drug and liquid chromatography/tandem mass spectrometry (LC-MS/MS) was used for quantitation. Post-dialysis
Tânia M G Almeida et al.
Chemistry & biodiversity, 2(12), 1691-1700 (2006-12-29)
Absorption, distribution, metabolism, and excretion (ADME) properties are of invaluable importance to a bioactive compound. Permeation process is one of the most widely studied by many different techniques. Among them, reversed-phase liquid chromatography (RP-LC) is proving to be of great
W Pan et al.
Journal of chromatography. B, Biomedical sciences and applications, 703(1-2), 129-138 (1998-02-03)
A sensitive, specific and precise HPLC-UV assay was developed to quantitate cocaine (COC) and its metabolites benzoylecgonine (BE), norcocaine (NC) and cocaethylene (CE) in rat plasma. After adding 50 microl of the internal standard solution (bupivacaine, 8 microg/ml) and 500
P Chaimbault et al.
Journal of chromatography. A, 855(1), 191-202 (1999-10-09)
A qualitative determination of 20 underivatized proteinic amino acids by LC-MS is reported. The need for chromatographic separation before mass spectrometry determination is demonstrated based on the study of several amino acid pairs which have some similar characteristics. Two suitable
Nasser E Bastani et al.
Rapid communications in mass spectrometry : RCM, 23(18), 2885-2890 (2009-08-12)
F2-isoprostanes are a family of prostaglandin F2-like compounds that are formed by free-radical-catalyzed peroxidation of arachidonic acid. Several F2-isoprostanes, but in particular 8-epi PGF(2alpha), are widely used as oxidative stress biomarkers. An analytical method based on liquid chromatography with negative

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service