Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

59191

Supelco

SUPELCOSIL ABZ+Plus HPLC Column

3 μm particle size, L × I.D. 3.3 cm × 4.6 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

agency

suitable for USP L60

feature

endcapped

manufacturer/tradename

SUPELCOSIL

extent of labeling

12.0% carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

3.3 cm × 4.6 mm

surface coverage

surface coverage 3.4 μmol/m2

matrix

silica gel, spherical particle platform

matrix active group

amide, alkyl phase

particle size

3 μm

pore size

120 Å

pH range

2-7.5

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

SUPELCOSIL ABZ+Plus columns offer both high deactivation and unique selectivity. Deactivated silica particles of very narrow particle size distribution ensure high efficiency with low back pressure. After bonding and endcapping reactions, the ABZ+Plus phase effectively shields unreacted silanol groups on the silica, preventing them from interacting with most analytes, and provides symmetric peaks regardless of an analyte′s functionality. The phase also allows you to use low ionic strength buffers without having to add an ion-suppressing modifier. ABZ+Plus enables you to use simple mobile phases when analyzing the most difficult compounds; acids, strongly basic compounds, and zwitterions.
suitable for L60 per USP

Features and Benefits

• High efficiency for polar, nonpolar, and charged analytes
• Symmetric peaks for the most reactive compounds
• Unique selectivity for polar and charged compounds

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Konstantinos Petritis et al.
Journal of chromatography. A, 957(2), 173-185 (2002-07-13)
The single run analysis of 23 small peptides (principally glycyl and lysyl dipeptides) is performed by ion-pair reversed-phase liquid chromatography coupled with evaporative light scattering detection or electrospray (tandem) mass spectrometry. Several perfluorinated carboxylic acid homologues are evaluated with an
Cinzia Stella et al.
Journal of separation science, 28(17), 2350-2362 (2005-12-14)
An RPLC was developed to rapidly determine lipophilicity of neutral and basic compounds using three base deactivated RPLC stationary phases particularly designed for the analysis of basic compounds, namely, Supelcosil ABZ(+)Plus, Discovery RP Amide C16, and Zorbax Extend C18. The
A B Baranda et al.
Talanta, 67(5), 933-941 (2008-10-31)
A liquid-liquid extraction method using diethyl ether as organic solvent was optimized simultaneously for five 1,4-dihydropyridines (amlodipine, nitrendipine, felodipine, lacidipine and lercanidipine) belonging to the group of calcium channel blockers. Some experimental tools such as a full factorial design, a
P Chaimbault et al.
Journal of chromatography. A, 855(1), 191-202 (1999-10-09)
A qualitative determination of 20 underivatized proteinic amino acids by LC-MS is reported. The need for chromatographic separation before mass spectrometry determination is demonstrated based on the study of several amino acid pairs which have some similar characteristics. Two suitable
Nasser E Bastani et al.
Rapid communications in mass spectrometry : RCM, 23(18), 2885-2890 (2009-08-12)
F2-isoprostanes are a family of prostaglandin F2-like compounds that are formed by free-radical-catalyzed peroxidation of arachidonic acid. Several F2-isoprostanes, but in particular 8-epi PGF(2alpha), are widely used as oxidative stress biomarkers. An analytical method based on liquid chromatography with negative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service