Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

59158

Supelco

SUPELCOSIL LC-F (5 µm) HPLC Columns

L × I.D. 25 cm × 4.6 mm, HPLC Column

Synonym(s):

SUPELCOSIL LC-F 5UM 25CMX4.6MM HPLC COLUMN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
Product 59158 is not currently sold in your country. Contact Technical Service

Product Name

SUPELCOSIL LC-F HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm

agency

suitable for USP L43

feature

endcapped

manufacturer/tradename

SUPELCOSIL

extent of labeling

5% Carbon loading

parameter

≤70 °C temp. limit
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.6 mm

surface area

170 m2/g

surface coverage

surface coverage 2.6 μmol/m2

matrix

silica gel, spherical particle platform

matrix active group

fluoro, penta- phase, amido

particle size

5 μm

pore size

120 Å

pH range

2-7.5

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

SUPELCOSIL LC-F columns contain a pentafluorophenyl functional group/endcapped packing material. These columns offer selectivities different from traditional reversed-phase columns for halogenated compounds, esters, ketones, bases, and taxanes, including taxol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jolanta Flieger et al.
Journal of separation science, 35(2), 248-255 (2012-01-01)
Sodium hexafluorophosphate, perchlorate and tetrafluoroborate were applied as the ion-pair reagents in reversed-phase chromatography of several imidazolium-based ionic liquids. The optimization of the retention was performed by changing the kind of organic modifier (methanol, acetonitrile), concentration and the kind of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service