Key Documents
59031
Column End Plugs
for 10-32 coned ports, red Delrin®, pkg of 10 ea
About This Item
UNSPSC Code:
41121815
material
red Delrin®
packaging
pkg of 10 ea
fitting
10-32 male
General description
Use column end plugs with all female 10-32 compression type end fittings, compatible with any common HPLC solvent.
Legal Information
Delrin is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)