Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

58969

Supelco

SUPELCOSIL LC-18-S (5 µm) HPLC Columns

L × I.D. 25 cm × 10 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

Product Name

SUPELCOSIL LC-18-S, L × I.D. 25 cm × 10 mm

technique(s)

HPLC: suitable

L × I.D.

25 cm × 10 mm

matrix

SUPELCOSIL silica gel

matrix active group

C18 (octadecyl) phase

diameter

5 μm

pore size

120 Å

application(s)

food and beverages

separation technique

reversed phase

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service