Key Documents
58826
Rheodyne® Model 7125 Injector
About This Item
UNSPSC Code:
41115854
feature
number of ports 6
positions 2
parameter
7000 psi max. pressure
General description
Rheodyne® Model 7125 injector comprises of a six-port sample injection valve in which the sample is loaded through a built-in needle port in front of the valve.
The Rheodyne Model 7125 syringe loading injector allows injection of the entire contents of the syringe into the sample loop -- you will not have to flush the valve between injections, unless you are conducting trace analyses. It also injects samples from a partially filled loop (you save time by not having to change the sample loop).
The Model 7125 injector can be used at pressures to 7000 psi (490 kg/cm2) and is supplied with a Vespel rotor seal for operation at pH 0-10. A 20 μL sample loop is included; order additional loops separately.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Rheodyne is a registered trademark of IDEX Health & Science, LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)