Key Documents
58789
SSI Two-Way Shut Off Valve
About This Item
UNSPSC Code:
41115854
description
two-way
feature
number of ports 2
General description
For 1/16 in. tubing; 1/4-28 (SSI style) fittings included. Rated to 15,000 psi (1054 kg/cm2).
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
SSI is a trademark of Scientific Systems, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)