Key Documents
58748
Glass-to-PTFE Connector
Joins PTFE tubing to glassware without coupling.
About This Item
UNSPSC Code:
41121815
material
acetal (polypropylene) union
glass tubing (flanged nipple)
packaging
pkg of 1 unit
glass tube L × O.D. × I.D.
3 1/4 in. × 1/4 in. × 2 mm
fitting
1/4-28 female UNF
General description
Glass-to-PTFE connector is highly advantageous in column chromatography as it helps in supplying solvents under pressure, gradient elution and degassing of solvents.[1] This device is safe and convenient to use to avoid scaling rings or septa damage from rubber connectors.[2]
Application
Glass-to-PTFE connector may be used for LPLC, TLC, paper chromatography and flash chromatography.
Legal Information
Valco is a registered trademark of Valco Instruments Co. Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Convenient apparatus for the generation of small amounts of diazomethane.
Cohen, Jerry D.
Journal of Chromatography A, 303, 193-196 (1984)
PTFE-glass pump. Chromatographic applications.
Arcus, A. C.
Journal of Chromatography A, 3, 411-414 (1960)