Key Documents
58744
Tubing Adapter
1/4-28 UNF for 1/16 in. O.D. tubing (flangeless fitting included), 10-32 Valco for 1/16 in. O.D. tubing (ZDV fitting included)
About This Item
UNSPSC Code:
41115700
material
316 stainless steel unit
description
for chromatography applications
packaging
pkg of 1 ea
bore
0.75 mm (0.031 in.)
fitting
1/4-28 UNF for 1/16 in. O.D. tubing (flangeless fitting included)
10-32 Valco for 1/16 in. O.D. tubing (ZDV fitting included)
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Valco is a registered trademark of Valco Instruments Co. Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)