Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

58690-U

SSI Prime/Purge Valve

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
SB.51

feature

number of ports 3

General description

The drain port in this valve is incorporated into the valve stem, eliminating the need for extra tubing and fittings. A luer taper on the valve stem accepts a luer syringe for priming. Valve shutoff is provided by a soft-seal tip that withstands back pressures to 8,000 psi (560 kg/cm2). Only PTFE, Tefzel, and 316 stainless steel come in contact with the mobile phase. Order 1/16 in. SSI nuts and ferrules separately.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SSI is a trademark of Scientific Systems, Inc.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service