Key Documents
58647
Ferrule Saver Tool
About This Item
UNSPSC Code:
41115700
General description
  • Quickly and simply dislodges swaged ferrule from 1/16 in. O.D. (0.010-0.030 in. I.D.) tubing
  • Saves time, money, and needless aggravation
Removes high pressure CPI-type stainless steel ferrules without the need for disassembling your HPLC system plumbing or cutting the tubing. Eliminates the need for opening, filing, and passivating tubing before installing a new ferrule. Opens and permits reuse of the removed ferrule. Not recommended for self-locking SSI and Rheodyne ferrules (could damage ferrule or tool).
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)