Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

58640-U

Upchurch Fittings Kit

for use with Bio-Rad/Rheodyne

Synonym(s):

UPTIGHT FITTINGS KIT,BIORAD SYSTEM/Rheodyne® VALVE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

compatibility

for use with Bio-Rad/Rheodyne

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

; S.S. ZDV Union, 0.050", 2 each ; Fingertight I Male Nut (one piece), 2 each ; S.S. ZDV Union, 0.020", 2 each ; Rheodyne Short Male Nut, 4 each ; Rheodyne Long Male Nut, 2 each ; Rheodyne Extra-Long Male Nut, 2 each ; Rheodyne Ferrule, 10 each ; S.S. Male Nut, 5 each ; S.S. Female Nut, 5 each ; S.S. Ferrule, 10 each ; Tefzel Cap, 2 each ; Delrin Plug, 2 each ; Stainless Steel Tubing, 5cm × 0.01" I.D., 4 each ; Stainless Steel Tubing, 20cm × 0.01" I.D., 4 each ; Stainless Steel Tubing, 5cm × 0.02" I.D., 4 each ; Stainless Steel Tubing, 10cm × 0.02" I.D., 4 each ; Stainless Steel Tubing, 20cm × 0.02" I.D., 4 each ; Stainless Steel Tubing, 10cm × 0.01" I.D., 4 each

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Rheodyne is a registered trademark of IDEX Health & Science, LLC

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service