Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

58635-U

Upchurch Fittings Kit

new

for use with Agilent/HP

Synonym(s):

UPTIGHT FITTINGS KIT,Hewlett-Packard SYSTEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

compatibility

for use with Agilent/HP

Looking for similar products? Visit Product Comparison Guide

Components

S.S. Ferrule, 20 eachStainless Steel Tubing, 10cm × 0.01" I.D., 4 eachStainless Steel Tubing, 20cm × 0.02" I.D., 4 eachStainless Steel Tubing, 10cm × 0.02" I.D., 4 eachSteel Tubing, 5cm × 0.02" I.D., 4 eachStainless Steel Tubing, 20cm × 0.01" I.D., 4 eachStainless Steel Tubing, 5cm × 0.01" I.D., 4 eachPEEK Replacement Ferrule, 4 eachFingertight II Male Nut (Delrin), 2 eachDelrin Plug, 2 eachTefzel Cap, 2 eachS.S. Female Nut, 10 eachS.S. Male Nut, 10 eachS.S. ZDV Union, 0.020", 2 each

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Hewlett-Packard is a trademark of Hewlett-Packard Corp.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service