Key Documents
58624
Supelco
Pressure Gauge Kits
parameter 0-6000 psi (0-420 bar)
About This Item
UNSPSC Code:
41115700
parameter
0-6000 psi (0-420 bar)
fitting
10-32 fittings
General description
Components already assembled, ready for easy installation. - Backed by a three-year warranty, these highly accurate (±1.5%), glycerine-filled gauges have a 2-1/2 in./6.4 cm face and 1/4 in. NPT connection. To resist corrosion, the one-piece case and socket and the bourdon tube are made of 316 stainless steel. Stainless steel tee with 1/16 inch fittings is included.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)