Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

58623-U

Supelco

Pressure Gauge Kits

parameter 0-3000 psi (0-210 bar)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

parameter

0-3000 psi (0-210 bar)

fitting

1/4 in. NPT gauge

Looking for similar products? Visit Product Comparison Guide

General description

Components already assembled, ready for easy installation. - Backed by a three-year warranty, these highly accurate (±1.5%), glycerine-filled gauges have a 2-1/2 in./6.4 cm face and 1/4 in. NPT connection. To resist corrosion, the one-piece case and socket and the bourdon tube are made of 316 stainless steel. Stainless steel tee with 1/16 inch fittings is included.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service