Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

58457AST

Supelco

CHIRALPAK® HSA (5 μm) HPLC Columns

L × I.D. 15 cm × 10 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

CHIRALPAK® HSA HPLC Column, 5 μm particle size, L × I.D. 15 cm × 10 mm

material

stainless steel column

agency

suitable for USP L79

product line

CHIRALPAK®

packaging

pkg of 1 ea

manufacturer/tradename

CHIRALPAK®

parameter

137 bar pressure (2000 psi)
20-30 °C temperature
40 °C max. temp.

technique(s)

HPLC: suitable

L × I.D.

15 cm × 10 mm

matrix

fully porous particle

matrix active group

human serum albumin phase

particle size

5 μm

operating pH

2-8

separation technique

chiral

Looking for similar products? Visit Product Comparison Guide

General description

CHIRALPAK HSA, which uses human serum albumin as the chiral selector, is highly selective for acidic racemates, preferably weak and strong acids, zwitterionic and non-protolytic (neutral) compounds. Phosphate buffers (normally 0.01-0.1M, pH 5-7) with addition of organic modifiers are used as mobile phases. Enantioselectivity and retention can be regulated by changing the mobile phase composition. However, the primary use of CHIRALPAK HSA is for fast drug/protein binding studies (1). To calculate the % protein binding, measure the retention time of an unretained compound (t0) and the compound of interest (tr) on the CHIRALPAK HSA column. Then use the capacity factor equation:
k = (tr - t0)/tr
to calculate the % protein binding (P):
P = 100k/(k+1)

Different types of mobile phases can be used. A mobile phase consisting of 6% 2-propanol in 20 mM potassium phosphate buffer, pH 7.0 gives data in good agreement with literature data. The mobile phase conditions should be chosen to suit the drugs to be tested, i.e., for high protein binding drugs a mobile phase with higher eluting strength might be needed in order to reduce retention times.

(1) Goodman, A.; Gilman, A.G. The Pharmacological Basis of Therapeutics, 9th Edition, McGraw-Hill: New York, 1996; pp 1712-1792.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

CHIRALPAK is a registered trademark of Daicel Corp.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service