A pulse damper controls pump pulsations for a more stable baseline. The LO-Pulse damper is a patented unit compatible with single piston reciprocating HPLC pumps (Altex 110A, Eldex pumps, LDC Mini-Pump VS, SSI Models 200 and 300, etc.). At pressures from 500 psi to 6,000 psi (35-420 kg/cm2), it improves precision of quantitative analyses and detection limits for trace sample components. Fittings and instructions included.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
LO-Pulse is a trademark of Scientific Systems, Inc.
SSI is a trademark of Scientific Systems, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)