Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

58410

Supelco

Supelco Silica and C18 Saturator HPLC Columns

Silica Saturator Column Kit

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41115700

Product Name

Supelco® Silica and C18 Saturator Columns, Silica Saturator Column Kit

agency

suitable for USP L1

feature

frits included

packaging

pkg of 1 kit

technique(s)

HPLC: suitable

matrix active group

C18 (octadecyl) phase

fitting

10-32 female Supelco housing for 1/16 in. O.D. tubing (nuts and ferrules included)

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

To protect your column from alkaline mobile phases which can dissolve silica (pH >7.0), saturate the mobile phase with silica by passing it through a silica saturator column.
To protect your column from acidic mobile phases which can strip bonded phase (pH <2.0), use a C18 saturator column.
Each kit consists of 10 g of packing (12 μm spherical particles, 180 m2/g surface area), a 7.5 cm × 4.6 mm column, two frits, and fittings to connect the column to 1/16 in. tubing.
The optional plastic funnel is helpful for filling these columns (Cat. No. 20390-U).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service