Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

58319

Supelco

Guard Column Hardware Kit

L × I.D. 5 cm × 4.6 mm

Synonym(s):

Postcolumn Reactor Components, Glass Beads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

material

stainless steel tubing

L × I.D.

5 cm × 4.6 mm

tubing L × O.D. × I.D.

5 cm × 1/16 in. × 0.01 in.

pore size

2.0 μm

Looking for similar products? Visit Product Comparison Guide

General description

Each Guard Column Hardware Kit contains an empty 5 cm x 4.6 mm ID column, 10 g of Pelliguard packing, 10 frits, and hardware for connecting the column to 1/16″ tubing. About 1.3 g of packing is needed to pack one 5 cm x 4.6 mm column.

Components

1 Column, 2 Frits, 1Tubing

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service