Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

58279-U

Valco® Pre-Column Filter

Screen Filter, pore size 2.0 μm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

material

stainless steel screen filter and fittings

packaging

pkg of 1 ea

pore size

2.0 μm

fitting

10-32 female Valco in-line for 1/16 in. O.D. tubing (fittings included)

Looking for similar products? Visit Product Comparison Guide

General description

In-line installation. Efficient, low dead volume filters protect your columns from particles without reducing column performance. The replaceable 1/8 in. frit has 0.5 μm pores to protect 3 μm or 5 μm column packings, the replaceable screen has 2 μm pores. Choose the frit filter for higher filtration capacity (most applications) or the screen filter for less dead volume (e.g., with microbore columns). Use with 1/16 in. O.D. tubing; 1/16 in. fittings included.

Other Notes

Frits and screens should not be interchanged in these filters.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Valco is a registered trademark of Valco Instruments Co. Inc.

replacement part

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service