Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

58162AST

Supelco

HPLC Column Coupler

PEEK, I.D. × O.D. × Overall L 0.007 in. × 1/16 in. × 1 in.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

material

PEEK

packaging

pkg of 1 ea

technique(s)

HPLC: suitable

I.D. × O.D. × Overall L

0.007 in. × 1/16 in. × 1 in.

fitting

10-32 male (Valco compatible)

compatibility

for use with HPLC columns with 1/16", 10-32 thread end-fittings (Used with all Supelco and Astec columns, plus other brands that have the same thread dimensions.)

General description

This guard column coupler may be used with all Astec® and CHIRALPAK® AGP, CBH, and HSA columns of 2 or 3 mm I.D. For 4 mm I.D columns, use coupler 54986.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALPAK is a registered trademark of Daicel Corp.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service