Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

58159AST

Supelco

Guard Column Holder for CHIRALPAK® AGP, HSA, and CBH

for use with CHIRALPAK AGP, CBH, and HSA 1 cm x 2.0, 3.0, and 4.0 mm guard cartridges

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

material

stainless steel

product line

CHIRALPAK®

packaging

pkg of 1 ea

technique(s)

HPLC: suitable

compatibility

for use with CHIRALPAK AGP, CBH, and HSA 1 cm x 2.0, 3.0, and 4.0 mm guard cartridges

General description

Guard column holder for CHIRALPAK AGP, CBH, and HSA 1 cm x 2.0, 3.0 and 4.0 mm I.D. guard cartridges

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

CHIRALPAK is a registered trademark of Daicel Corp.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service