Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

58094

Aura Mobile Phase Filter/Degasser Unit

reservoir size 1 L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

reservoir size

1 L

Looking for similar products? Visit Product Comparison Guide

General description

Easy-to-use Aura filter/degasser units simultaneously degas solvents and buffers and remove particles. You can eliminate spurious peaks caused by gas bubbles in your detector, and prevent particle damage to check valves and other system components. A clean mobile phase also prolongs column life.

Each unit includes a PTFE filtering/degassing assembly, ten 1.5 μm PTFE filter membranes, 40 in./1 m × 1/4 in. O.D. PTFE tubing, and a heavy wall, graduated, borosilicate glass solvent reservoir. Choose either a 1-liter or a 2-liter reservoir. The standard taper PTFE joint on the filtering assembly fits into Erlenmeyer flasks and other containers. The filtering assembly accepts any standard 47 mm filter membrane and is compatible with all solvents used in HPLC. Use with appropriate glassware and any vacuum pump.

The Aura filter/degasser is particularly suitable for use with solid phase extraction disks (SPED). It ensures simple disc placement, large filtration area, small restriction to flow from the disk support element, and the convenience and advantages of the liquid inlet tube (allows sediments to be transferred last).

Attaches to any vacuum line.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. What filters can I use with the Aura Mobile Phase Filter/Degasser Unit, Product No. 58094?

    1 answer
    1. The direct PTFE replacement filters for this filter/degasser unit is Product No. 58086.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service