Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

57920

Supelco

SUPELCOSIL LC-18-S (5 µm) HPLC Columns

L × I.D. 30 cm × 1 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41115700
eCl@ss:
32110501

Product Name

SUPELCOSIL LC-18-S HPLC Column, 5 μm particle size, L × I.D. 30 cm × 1 mm

agency

suitable for USP L1

feature

endcapped

manufacturer/tradename

SUPELCOSIL

extent of labeling

11.0% carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

30 cm × 1 mm

surface area

170 m2/g

surface coverage

surface coverage 3.1 μmol/m2

matrix

silica gel, spherical particle platform

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

120 Å

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

SUPELCOSIL LC-18-S columns are designed for reliable separations of deoxyribonucleosides and ribonucleosides.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mert Ilhan et al.
Evidence-based complementary and alternative medicine : eCAM, 2014, 578103-578103 (2014-08-29)
The aim of the present research is to evaluate the antiurolithiatic effect of the various extracts prepared from the fruits of Viburnum opulus L., in regard to its ethnobotanical record. To induce urolithiasis, 70 mg/kg sodium oxalate was injected to the
L Marai et al.
Canadian journal of biochemistry and cell biology = Revue canadienne de biochimie et biologie cellulaire, 61(8), 840-849 (1983-08-01)
Natural triacylglycerols were resolved by high pressure liquid chromatography on Supelcosil LC-18 columns using a gradient of 30-90% propionitrile in acetonitrile as eluting solvent. The effluent was admitted to a quadrupole mass spectrometer via a direct liquid inlet interface. The
V Stocchi et al.
Analytical biochemistry, 146(1), 118-124 (1985-04-01)
A simple and rapid method for the determination of ATP, ADP, AMP, NADP+, NAD+, NADPH, and NADH in human erythrocytes is described. A single-step extraction procedure employing alkaline medium and CF 50A Amicon ultrafiltration membranes allows a simultaneous and total
Catrina L Dupard-Julien et al.
Analytical and bioanalytical chemistry, 387(3), 1027-1032 (2007-01-06)
A reversed-phase, high-performance liquid chromatography (RP-HPLC) method that allows quantitation of low levels of epoxides has been described. The method involved derivatization of epoxides using 100- to 1,000-fold excess N,N-diethyldithiocarbamate (DTC) at 60 degrees C for 20 min at neutral
K M Matar et al.
Therapeutic drug monitoring, 21(5), 559-566 (1999-10-16)
A simple, rapid, sensitive, and reproducible high-performance liquid chromatographic (HPLC) method for simultaneous determination of the antiepileptic drugs (ethosuximide, primidone, lamotrigine, phenobarbital, phenytoin, and carbamazepine) and two metabolites (carbamazepine-diol and carbamazepine-epoxide) in human plasma is described. The procedure involves extraction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service