Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

57652-U

Upchurch PEEK LiteTouch® Fitting

PEEK with stainless steel lock ring ferrule (for 1/8″ tubing)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

material

PEEK with stainless steel lock ring ferrule (for 1/8″ tubing)

packaging

pkg of 10 ea

parameter

4500 psi max. pressure (310 bar)

fitting

Litetouch fitting Upchurch ferrule for 1/8 in. O.D. tubing

Looking for similar products? Visit Product Comparison Guide

General description

Unique design prevents twisting of PEEK tubing. The fitting consists of a special PEEK ferrule, a stainless steel ring, and a PEEK or stainless steel nut for 1/16 in. tubing. The fitting will not leak at pressures to 5000 psi/340 bar with the PEEK nut, 2500 psi/175 bar with the stainless steel nut finger-tightened or 7000 psi/480 bar with wrench tightening. After wrench-tightening the stainless steel nut, subsequent finger-tightening will hold to 5000 psi. Compatible with all manufacturers′ stainless steel nuts. Order nuts and ferrules separately.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiteTouch is a registered trademark of IDEX Health & Science, LLC

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service