Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

57629-U

Sample Loop for Model 7725 Injector

volume 500 μL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
eCl@ss:
42079206
NACRES:
SB.51

material

stainless steel sample loop and fittings

volume

500 μL

compatibility

for use with Rheodyne 7725 and 7725i injector

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service