Key Documents
57622
Sample Loop for 9725 Injector
volume 2 μL
About This Item
UNSPSC Code:
41115854
volume
2 μL
Other Notes
Note: Use VESPEL seals to pH 10, Tefzel seals to pH 14.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)