Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

575680-U

Supelco

96 Round Well Pierceable Cap Mats

for use with HybridSPE-PLus 96-well plates, pk of 50 ×

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
NB.21

packaging

pk of 50 ×

technique(s)

solid phase extraction (SPE): suitable

application(s)

food and beverages

compatibility

for use with HybridSPE-PLus 96-well plates

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Novel sample prep technique coupled with the unique selectivity of the Ascentis Express F5 column enables a fast and simplified bioanalytical method for associated vitamin D metabolites.

Using HybridSPE-Phospholipid, Ascentis Express, and Other LC-MS Workflow Components

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service