Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

575611-U

Supelco

Discovery® DSC-PS/DVB SPE 96-well Plate

bed wt. 50 mg/well, pkg of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104018

composition

bed wt., 50 mg/well

packaging

pkg of 1 ea

technique(s)

solid phase extraction (SPE): suitable

surface area

800-950 m2/g

matrix

DSC-PS/DVB styrene-divinylbenzene (non-ionic)

matrix active group

polystyrene-divinylbenzene bonding

diameter

80-160 μm

pore size

110-175 Å

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service