Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

57484-U

Supelco

LiChrosorb® RP-8 (5 µm) HPLC Columns

L × I.D. 12.5 cm × 4 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

LiChrosorb® C8 12.5CM x 4.1MM HPLC COLUMN, column L × I.D. 12.5 cm × 4 mm

material

stainless steel column

agency

suitable for USP L7

product line

LiChrosorb®

feature

endcapped: no

manufacturer/tradename

LiChrosorb®

packaging

1 ea of

parameter

0-70 °C temperature
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

column L × I.D.

12.5 cm × 4 mm

matrix

silica particle platform
fully porous particle

matrix active group

C8 (octyl) phase

diameter

5 μm

pore size

100 Å pore size

operating pH

2-7.5

separation technique

reversed phase

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChrosorb is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service