Key Documents
57454-U
Supelco
SPME PTFE Sealing Caps
for use with 23 GA fibers, pk of 3
About This Item
UNSPSC Code:
41115854
NACRES:
NB.21
material
PTFE fiber
packaging
pk of 3
technique(s)
solid phase microextraction (SPME): suitable
application(s)
food and beverages
compatibility
for use with 23 GA fibers
General description
Sealing caps protect the SPME fiber assembly from accidental damage to the needle tip and from contamination by dust and dirt. The caps also provide an airtight seal which protects the 23 gauge SPME fibers from contamination or loss of analytes when using adsorbent fiber coatings.
Choose from one of the most recent versions:
Certificates of Analysis (COA)