Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

57389-U

SPME Fiber Assembly Storage Device

for use with SPME Fiber Assemblies

Synonym(s):

SPME Storage Devices

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

compatibility

for use with SPME Fiber Assemblies

Looking for similar products? Visit Product Comparison Guide

General description

Devices for safely storing conventional SPME fiber assemblies or SPME Fast Fit Assemblies (FFA) to maintain fiber conditioning prior to sampling and sample integrity after sampling. The storage containers are ideal for shipping fibers to and from sampling sites or just to keep them clean and ready for sampling in the laboratory.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esther De La Fuente et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 44(10), 1004-1010 (2009-10-16)
The complex molecular assemblages were analysed in the soil gas phase after applying pine forest wastes (PFW) or sugarbeet vinasses (SBV) for soil-borne crop pests' management. For this purpose, solid-phase micro-extraction (SPME) and gas chromatography (GC) were used coupled with
J Tytgat et al.
International journal of legal medicine, 109(3), 150-154 (1996-01-01)
The most common alkyl nitrites encountered in forensic toxicology are iso-butyl, n-butyl and iso-pentyl(amyl) nitrites. All have become popular as an aphrodisiac, especially among the homosexual population. Alkyl nitrites are a volatile and unstable group of compounds, which hydrolyse in
Jermiah Y Shen et al.
Journal of separation science, 35(19), 2544-2552 (2012-08-22)
The presence of triclosan and triclocarban, two endocrine-disrupting chemicals and antimicrobial agents, and transformation products of triclocarban, 1,3-di(phenyl)urea, 1,3-bis(4-chlorophenyl)urea and 1,3-bis(3,4-dichlorophenyl)urea, in tap water, treated household drinking water, bottled water, and river water samples were investigated using solid-phase micro-extraction coupled
Marghereth Saverini et al.
Journal of photochemistry and photobiology. B, Biology, 108, 8-15 (2012-01-20)
The presence of (±)α-pinene, (+)β-pinene, (+)3-carene, and R-(+)limonene terpenes in wastewater of a citrus transformation factory was detected and analyzed, in a previous study, by using Solid Phase Micro-extraction (SPME) followed by GC analyses. Purpose of that research was to
Y Bezman et al.
Journal of agricultural and food chemistry, 49(11), 5425-5432 (2001-11-21)
The occurrence of methional in fresh orange juice, and possible occurrence of beta-damascenone in heated orange juice, has been previously suggested. Here we report on the occurrence of 2-methyl-3-furanthiol in the headspace, collected by solid-phase micro-extraction, of fresh, pasteurized, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service