When used with a standard 47mm glass filtration apparatus, the ENVI-Disk Clamp creates a better seal, eliminating leaks with SPE extraction disks or when filtering HPLC mobile phase solvents.
Use only with a filtration glassware funnel base that has a removable filtration stage, such as Supelco Mobile Phase Filtration Apparatus 1 (Cat. No. 58061) or 2 (Cat. No. 58062-U), or with a funnel base (Cat. No. 58064 or 58068). It cannot be used with a permanent fritted glass filtration stage or stainless steel holder screen.
Features and Benefits
Eliminates leaks
Attaches to any 34/45 tapered flasks
Legal Information
ENVI-Disk is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)