Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

57214-U

Supelco

Discovery® DSC-SAX SPE Bulk Packing

pkg of 100 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

packaging

pkg of 100 g

technique(s)

solid phase extraction (SPE): suitable

matrix active group

ammonium ion, quaternary bonding

application(s)

food and beverages

separation technique

ion exchange

General description

Retention Mechanism: Anion-exchange
Sample Matrix Compatibility: Organic or aqueous solutions

  • A polymerically bonded quarternary amine that remains positively charged at all pH levels
  • Counter ion is Cl-
  • Ion exchange capacity is ∼ 0.14 meq/g
  • Commonly used when extracting weaker anions (e.g., carboxylic acids) that may not bind strongly enough to weaker anion-exchangers
  • Selectivity can be modified by changing the counter ion with the appropriate buffer during conditioning
·

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Normal-phase SPE separates analytes based on polar interaction with sorbents in diverse sample matrices.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service