Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

57206

Supelco

Supelclean LC-Alumina A SPE Bulk Packing

pkg of 100 g

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41115712

packaging

pkg of 100 g

technique(s)

solid phase extraction (SPE): suitable

matrix

LC-Alumina-A Alumina for acidic pH (~5); Brockman Act. I

matrix active group

Alumina phase

diameter

60-325 mesh

application(s)

food and beverages

separation technique

normal phase

Legal Information

Supelclean is a trademark of Sigma-Aldrich Co. LLC

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service