Supelclean SPE Method Development Kits consist of an assortment of SPE phase chemistries and cartridge configurations ideal for SPE method development. The range of phase chemistries available for each kit allows the user to profile for compound retention, elution and sample matrix selectivity.
Components
Kit contains 6 of each phase chemistry:
Supelclean LC-Si, 500mg/3mL
Supelclean LC-Diol, 500mg/3mL
Supelclean LC-NH2, 500mg/3mL
Supelclean LC-Alumina-A, 1g/3mL
Supelclean LC-Alumina-N, 1g/3mL
Supelclean LC-Alumina-B, 1g/3mL
Legal Information
Supelclean is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)