Key Documents
56768
Eppendorf® CH-430 column heater
AC/DC input 120 V AC
About This Item
UNSPSC Code:
41115854
Features and Benefits
The model CH-430 column heater is like the model CH-30, except that it is wider, to accommodate up to four 30 cm long
columns (not including end fittings). The CH-430 heater also has provisions for heating two 7000 series Rheodyne valves.
Other Notes
No CE mark
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)