Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

56760-U

Eppendorf® TC-45 temperature controller

AC/DC input 120 V AC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
SB.51

compatibility

for use with Eppendorf Oven

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

This low-cost controller will control the CH-30 Eppendorf heater from 30-65 °C. Temperature is set via DIP switches in 5 °C increments.

Other Notes

No CE mark.

Footnote

Does not have a CE mark

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service