Key Documents
56646-U
Supelco
HyperShear Ultra HPLC High Pressure Static Mixer
In-line housing assembly, volume 350 μL
About This Item
UNSPSC Code:
41115854
material
stainless steel
description
Port size, 10-32 CPI (standard HPLC fittngs for 1/16" tubing)
Pressure rated to 18,000 psi
wetted material 316 stainless steel/PEEK
volume
350 μL
General description
HyperShear Mixers incorporate a highly efficient cross-flow shearing mechanism which produces vortex shear mixing over a wide flow range. This mixing technology typically delivers between 25% to 200% better mixing efficiency compared to conventional packed bed or tortuous path mixers.
Choose from one of the most recent versions:
Certificates of Analysis (COA)