Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

56274-U

O-Ring for Omnifit® Column Endpiece

for use with 10mm Bore Columns, Viton®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
SB.52

material

Viton®

packaging

pkg of 10 ea

manufacturer/tradename

Omnifit 6466

technique(s)

LPLC: suitable

compatibility

for use with 10mm Bore Columns

Looking for similar products? Visit Product Comparison Guide

General description

O-ring sealing components are used in machines required to seal the fluids or gases.[1] Column end piece tend to trap liquid around the O-ring sealing, to avoid such problem the upper column end piece can be redesigned.[2]

Legal Information

Omnifit is a registered trademark of Diba Industries
Viton is a registered trademark of E. I. du Pont de Nemours and Company

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peptide synthesis. Part 13. Feedback control in solid phase synthesis. Use of fluorenylmethoxycarbonyl amino acid 3, 4-dihydro-4-oxo-1, 2, 3-benzotriazin-3-yl esters in a fully automated system.
Cameron, Linda R., et al.
Journal of the Chemical Society. Perkin Transactions 1, 10, 2895-2901 (1988)
Anthony Sofronas
Analytical Troubleshooting of Process Machinery and Pressure Vessels (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service