Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

56202AST

Supelco

apHera C8 (5 μm) HPLC Columns

L × I.D. 15 cm × 4.6 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

apHera C8 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm

material

stainless steel column

product line

apHera

packaging

pkg of 1 ea

manufacturer/tradename

apHera

parameter

147 bar pressure (2133 psi)
4-60 °C temperature

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

15 cm × 4.6 mm

matrix

polymeric

matrix active group

C8 (octyl) phase

particle size

5 μm

pore size

300 Å

operating pH

1-14

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

apHera reversed columns were devoloped specifically to provide the superior advantages of both silica and polystyrene columns, without the disadvantages of either. This was accomplished using a vinyl alcohol copolymer base that keeps the surface wetted even with high carbon loads. The porous structure has an average pore diameter large enough to produce ideal results for small analytes, peptides and small proteins. These columns equal silica based columns in separation efficiency with organic solvents but provide efficiency with buffered alkaline solutions not possible on silica. One of the most significant features is logicial elution order of alkylated bases where retention increases proportionately with increasing chain length.

Features of apHera Reversed Phase Columns
  • Stable vinyl copolymer base
  • Stable pH range 2-12
  • 300 angstrom pore size
  • Stable in all organic solvents
  • Expanded applications: peptides, proteins
  • Efficient separation of any basic substance
  • Amenability to washing and alkaline solutions

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

apHera is a trademark of Sigma-Aldrich Co. LLC

guard column


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service