Key Documents
56138
Omnifit® Small Bore Connector
Two-way connector
About This Item
UNSPSC Code:
12352200
description
PTFE cones included
manufacturer/tradename
Omnifit 2415
technique(s)
LPLC: suitable
bore
0.8 mm
General description
Omnifit® Bore Connector is very versatile in nature and is used for connecting various types of hardwall tubings. The bore connector has a manual valve operation on each end, thus the flow can be switched on or off as required. It is primarily used for fluid-line interconnection, sampling and mixing.[1]
Application
Omnifit® Bore Connector was used to construct the flow-injection system for amperometric determination of tacrine.[2]
Features and Benefits
PTFE cone included.
Legal Information
Omnifit is a registered trademark of Diba Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Flow-Injection Amperometric Determination of Tacrine based on Ion Transfer across a Water-Plasticized Polymeric Membrane Interface.
Joaquin, A. C.
Sensors, 7, 1185-1192 (2007)
Modern Instrumental Techniques
Keith, A
Soil and Environmental Analysis, 1-700 (2003)