56108
Frit for Omnifit® Column Endpiece
PTFE, for use with 10 mm bore, pore size 5 μm
About This Item
Recommended Products
material
PTFE
packaging
pkg of 20 ea
manufacturer/tradename
Omnifit 6147
technique(s)
LPLC: suitable
pore size
5 μm
compatibility
for use with 10 mm bore
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service