Key Documents
56100-U
Omnifit® Column Connector
About This Item
UNSPSC Code:
41115716
NACRES:
SB.52
manufacturer/tradename
Omnifit 6164
technique(s)
LPLC: suitable
Application
Joins 2 columns of same diameter, 50 psi max.
Legal Information
Omnifit is a registered trademark of Diba Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)