Key Documents
56085
Omnifit® Cap Adapter
male Luer
About This Item
UNSPSC Code:
12000000
manufacturer/tradename
Omnifit 1201
technique(s)
LPLC: suitable
fitting
male Luer
5/16 in. female BSF for Omnifit column endfitting (Omnifit cap)
General description
Omnifit® Cap Adapter is used to connect an Omnifit connector or column end fitting and Luer fitting or soft tubing.
Legal Information
Omnifit is a registered trademark of Diba Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)