Key Documents
56069
Omnifit® Tube Endfittings
for use with 1/16 in. O.D. tubing, white Tefzel®
About This Item
UNSPSC Code:
23151817
material
white Tefzel®
packaging
pkg of 10 ea
technique(s)
LPLC: suitable
fitting
1/4-28 male UNF nut (Gripper Fitting required)
compatibility
for use with 1/16 in. O.D. tubing
General description
Each endfitting has an internal taper to hold the tubing centrally; includes a lock nut for panel mounting.
Gripper fitting required (purchase separately):
for 1/16in. O.D. tubing p/n 57417
for 1/8in. O.D. tubing p/n 57418
Legal Information
Omnifit is a registered trademark of Diba Industries
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
Study of potential leakage on several stressed fittings for hydrogen pressures up to 700 bar.
Deborah, H. D.
International Journal of Hydrogen Energy, 37(22), 17509-17516 (2012)
Salvatore, F.
Liquid Chromatography: Fundamentals and Instrumentation (2013)
Interpreting the difference between conventional and bi-directional plate-height measurements in liquid chromatography
Hui, G. D.
Journal of Chromatography A, 1217(40), 6214-6229 (2010)
Liquid chromatography on an amide stationary phase with post-column derivatization and fluorimetric detection for the determination of streptomycin and dihydrostreptomycin in foods.
Pilar, V. N.
Talanta, 72(2), 808-812 (2007)
John W.D.
Troubleshooting LC Systems (1989)