Key Documents
56061
Omnifit® Tube Endfittings
for use with 1/16 in. O.D. tubing, orange polypropylene
About This Item
UNSPSC Code:
12352200
material
orange polypropylene
packaging
pkg of 10 ea
technique(s)
LPLC: suitable
fitting
1/4-28 male UNF nut (Gripper Fitting required)
compatibility
for use with 1/16 in. O.D. tubing
General description
Each endfitting has an internal taper to hold the tubing centrally; includes a lock nut for panel mounting.
Gripper fitting required (purchase separately):
for 1/16in. O.D. tubing p/n 57417
for 1/8in. O.D. tubing p/n 57418
Legal Information
Omnifit is a registered trademark of Diba Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
John W.D.
Troubleshooting LC Systems (1989)
Salvatore, F.
Liquid Chromatography: Fundamentals and Instrumentation (2013)
Study of potential leakage on several stressed fittings for hydrogen pressures up to 700 bar.
Deborah, H. D.
International Journal of Hydrogen Energy, 37(22), 17509-17516 (2012)
Liquid chromatography on an amide stationary phase with post-column derivatization and fluorimetric detection for the determination of streptomycin and dihydrostreptomycin in foods.
Pilar, V. N.
Talanta, 72(2), 808-812 (2007)
Interpreting the difference between conventional and bi-directional plate-height measurements in liquid chromatography
Hui, G. D.
Journal of Chromatography A, 1217(40), 6214-6229 (2010)