55326-U
Supel™ Genie HybridSPE® Online SPE Cartridges
L × I.D. 20 mm × 4.0 mm, pkg of 2
About This Item
product line
Supel™
composition
bed wt., 100 mg
packaging
pkg of 2
technique(s)
solid phase extraction (SPE): suitable
L × I.D.
20 mm × 4.0 mm
matrix active group
zirconia-based phase
application(s)
food and beverages
Looking for similar products? Visit Product Comparison Guide
General description
For first time use, this starter kit is required to obtain reusable universal hardware that will fit all Supel Genie Online SPE cartridges.
LC instrumentation must be configured to accommodate Online SPE cartridges. Please see instrument setup diagram in data sheet 55326-U
Please contact [email protected] for more information on setup and hardware.
Application
Features and Benefits
- Provides cleaner samples for longer column life, less instrument downtime, and more accurate and reproducible data
- Automation saves time, decreases costs, and eliminates human error
- Unique Lewis acid/base retention mechanism effectively remove >95% of phospholipids from both hydrophilic and hydrophobic analytes of interest (unlike competitive chemistries) eliminating phospholipid-induced ion suppression
- Highly reproducible, consistent product performance
- Starter kit comes with reusable universal hardware (fits any of the Supel Genie Online SPE cartridges) and 1 cartridge
- Additional cartridges are also available in packs of 2 or 6
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service